Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-p38α(Thr180/Tyr182) Rabbit anti-Human, Mouse, Vertebrates, Clone: 8.78.8, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 051059
Description
Anti-phospho-p38α(Thr180/Tyr182) Antibody, clone 8.78.8 detects level of phospho-p38α(Thr180/Tyr182) and has been published and validated for use in western blotting and immunofluoresence.Specifications
phospho-p38α(Thr180/Tyr182) | |
Monoclonal | |
Unconjugated | |
NP_001306 | |
Synthetic peptide corresponding to amino acids surrounding and including phosphorylated Thr180/Tyr182 of human p38α. | |
Primary | |
Human, Mouse, Vertebrates | |
Supernatant |
Immunofluorescence, Western Blot | |
8.78.8 | |
Rabbit Monoclonal antibody in culture supernatant containing 0.05% sodium azide. | |
Rabbit | |
100 μL | |
P38B, P38BETA2, PRKM11, SAPK2, SAPK2B, p38-2, p38Beta, p38b | |
−20°C for 12 months | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only