Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p53 (Ser15) Mouse anti-Human, PE, Clone: 261352, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523586

Catalog No. PIMA523586


Add to cart

Description

Description

In Western blots, no cross-reactivity with human p53 that is unphosphorylated at S15 is detected.

The tumor suppressor protein, p53, is a sequence specific transcription factor that is activated by cellular stress. p53 mediates cell cycle arrest or apoptosis in response to DNA damage or starvation for pyrimidine nucleotides. p53 is up-regulated in response to stress signals and stimulated to activate transcription of specific genes, resulting in expression of p21waf1 and other proteins involved in G1 or G2/M arrest. The structure of p53 comprises an N-terminal transactivation domain, a central DNA-binding domain, an oligomerisation domain, and a C-terminal regulatory domain. There are various phosphorylation sites on p53, of which the phosphorylation at Ser15 is important for p53 activation and stabilization. p53 has been characterized to play a role in blocking the proliferative action of damaged cells and act as an anticancer agent. Phosphorylation of Ser392 in p53 has been shown to associate with the formation of human tumors. In addition, p53 has also been linked to the effects of aging and oxidative stress and an increase in p53 has been linked to deficits in LTP (Long Term Potentiation) in learning and memory. p53 is found in very low levels in normal cells, however, in a variety of transformed cell lines, it is expressed in high amounts, and believed to contribute to transformation and maligcy. Mutants of p53 that frequently occur in a number of different human cancers fail to bind the consensus DNA binding site, and cause the loss of tumor suppressor activity. Alterations of the TP53 gene occur not only as somatic mutations in human maligcies, but also as germline mutations in some cancer-prone families such as Li-Fraumeni syndrome.
Specifications

Specifications

Phospho-p53 (Ser15)
Monoclonal
PE
TP53
Liquid
TP53
Phosphopeptide containing the human p53 S15 site
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
261352
Buffered saline solution with BSA and 0.09% sodium azide
P04637
antigen NY-CO-13, cellular tumor antigen p53, p53 tumor suppressor, phosphoprotein p53, transformation-related protein 53, TP53
Mouse
IgG1
100 Tests
4° C, store in dark
Primary
7157
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit