Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-p53 (Ser6) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA517893
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with p53 phosphorylated at other sites.
Phospho-p53 (Ser6) Polyclonal antibody specifically detects Phospho-p53 (Ser6) in Hamster, Human, Non-human Primate samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Phospho-p53 (Ser6) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O09185, P04637 | |
Tp53 | |
Synthetic phosphopeptide corresponding to residues surrounding pSer6 of human p53 | |
100 μL | |
Primary | |
Hamster, Human, Non-human Primate | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100682525, 7157 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction