Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phospho-p70 S6 Kinase pThr389 Clone: B2H9L2, Invitrogen™

Phospho-p70 S6 Kinase pThr389 Monoclonal antibody 701064 from Thermo Scientific™ Pierce™ specifically detects Phospho-p70 S6 Kinase pThr389 in Human samples, and it is validated for WB, IF and ICC

Manufacturer:  Invitrogen 701064

Encompass

Catalog No. 70-106-4


Add to cart

Description

Description

p70 Ribosomal Protein S6 Kinase (p70S6K) is a 70 kDa member of the ribosomal S6 kinase (RSK) family of serine/threonine kinases. It is predominantly localized in the cytoplasm, and is essential in growth factors regulated cell proliferation, pathways involving cell motility, such as metastases, the immune response, and tissue repair. p70S6K acts downstream of phosphoinositide (PI) 3-kinase, and its main physiological target is the S6 ribosomal protein, which is involved in upregulation of protein synthesis.

701064 detects Phospho-p70 S6 Kinase pThr389 in Human samples. 701064 has been successfully used in WB, IF and ICC procedures. The immunogen for 701064 is phosphopeptide corresponding to amino acids 384-394 of human p70 ribosomal protein S6 kinase.

Applications:

Immunocytochemistry, Immunofluorescence, Western Blotting

Specifications

Specifications

Phospho-p70 S6 Kinase pThr389
Unconjugated
P23443
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Monoclonal
Human
B2H9L2
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
Phosphopeptide corresponding to amino acids 384-394 of human p70 ribosomal protein S6 kinase
Protein A
Primary
6198
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit