Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p73 (Tyr99) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141357

Catalog No. 01-675-491


Add to cart

Description

Description

Suggested positive control: antigen standard for TP73 (transient overexpression lysate).

p73 was identified as a long-lost cousin of tumor suppressor protein, p53 (4). p73 has high homology with p53 as well as with p63, a gene implicated in the maintece of epithelial stem cells. Significant homology between p53, p63, and p73 (approximately 63% amino acid identity in the DNA-binding domain suggest that they may have overlapping functions in the regulation of gene expression (3). The targeted disruption of p73 gene leads to defects hippocampal dysgenesis, hydrocephalus, chronic inflammation and infections (2). Recently, spilicing variant mRNAs of p73 has been identified in MCF-7, a breast carcinoma cell line. These mRNAs code for variant p73 proteins bearing distinct carboxy-terminal structures suggesting that the carboxy-terminal region of p73 may be important for the functions of this protein (1).
Specifications

Specifications

Phospho-p73 (Tyr99)
Polyclonal
Unconjugated
TP73
Liquid
TP73
Synthetic phosphopeptide corresponding to residues 75-125 of human p73, containing a phosphorylated tyrosine residue at position 99.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.2 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
O15350
P73
Rabbit
IgG
20 μg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
7161
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit