Learn More
Phospho-PAK1/2/3 (Thr402) Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14636
Catalog No. PA14636
Add to cart
Description
Description
This antibody is predicted to react with bovine, canine, human and mouse based on 100% sequence homology. This antibody is specific for the ~68 kDa to ~70 kDa PAK phosphorylated at Thr402. Immunolabeling can be blocked by lambda-phosphatase treatment.
In mammals, there are several identified isoforms of p21-Activated Protein Kinases or PAKs: alpha-PAK (also known as PAK-1) and beta-PAK (also known as PAK-3) are mostly brain-specific, while gamma-PAK (also known as PAK-2) is expressed ubiquitously (Jakobi et al., 2003). Mutations of the gene coding for PAK-3 are associated wit X-linked mental retardation and recent work indicates that PAK-3 is a key regulator of synapse formation and plasticity in the hippocampus (Boda et al., 2004). PAK-3 is thought to play a key role in regulation of cell shape and motility as well as cell death (Jakobi et al., 2003; Walter et al., 1998). Autophosphorylation of Thr402 in the protein has been found to be essential for activation of PAK (Jakobi et al., 2000).
Specifications
Specifications
Phospho-PAK1/2/3 (Thr402)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P35465, Q62829, Q64303
Pak1, Pak2, Pak3, p21-Activated Protein Kinase-1, 2, 3, Serine/threonine-protein kinase PAK 1, Alpha-PAK, p21-activated kinase 1, PAK-1, p65-PAK, Serine/threonine-protein kinase PAK 2, Gamma-PAK, PAK65, S6/H4 kinase, p21-activated kinase 2, PAK-2, p58, PAK-2p27, PAK-2p34, p34, C-t-PAK2, Serine/threonine-protein kinase PAK 3, Beta-PAK, Oligophrenin-3, p21-activated kinase 3, PAK-3, OPHN3
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29431, 29432, 29433
Western Blot
Unconjugated
Pak3
Liquid
PAK1, PAK2, PAK3
Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr402 of rat PAK1-3 conjugated to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Rat