Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Paxillin (Tyr31) Mouse anti-Human, Clone: M102, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051143
Description
Detect phospho-Paxillin (Tyr31) using this Anti-phospho-Paxillin (Tyr31) Antibody, clone M102 validated for use in western blotting.Specifications
phospho-Paxillin (Tyr31) | |
Monoclonal | |
Unconjugated | |
P49023 | |
Mouse | |
Protein A chromatography | |
Primary | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Western Blot | |
M102 | |
Purified monoclonal IgG1 in phosphate-buffered saline, 50% glycerol, 1mg/mL BSA, and 0.05% sodium azide. | |
FLJ16691, paxillin | |
Clone M102 was generated from phospho-Paxillin (Tyr31) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 31 of human paxillin. This human sequence is highly conserved in rat and mouse paxillin. | |
100 μL | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only