Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PKA beta (Ser338), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126733

Catalog No. PIPA126733

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-26733 detects PKA beta (catalytic subunit) (phospho S338) from human and mouse samples. PA1-26733 is expected to cross react with porcine and bovine due to sequence homology.PA1-26733 has been successfully used in Western blot procedures.The PA1-26733 immunogen is a synthetic phosphopeptide corresponding to a portion of human PKA catalytic beta subunit containing serine 338.
Specifications

Specifications

Phospho-PKA beta pSer338
Polyclonal
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3
P05131, P22694, P68181, P05383
PRKACB
Synthetic phosphopeptide corresponding to a portion of human PKA catalytic beta subunit containing serine 338.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Murine, Porcine
Western Blot
Unconjugated
PRKACB
Purified
Rabbit
IgG
50μL
Store at -20°C
Primary
5567
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit