Learn More
Phospho-PKC beta-2 (Thr641) Rabbit anti-Human, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen 44964G
Catalog No. 44-964-G
Add to cart
Description
Description
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase has been reported to be involved in many different cellular functions, such as B cell activation, apoptosis induction, endothelial cell proliferation, and intestinal sugar absorption. Studies in mice also suggest that this kinase may also regulate neuronal functions and correlate fear-induced conflict behavior after stress. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
Specifications
Phospho-PKC beta-2 (Thr641)
Polyclonal
Dulbecco′s PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3
P05771
Protein kinase C beta type, PKC-B, PKC-beta, PRKCB, PKCB, PRKCB1
Rabbit
IgG
100 μL
-20°C
Primary
5579
Western Blot
Unconjugated
PRKCB
Liquid
PRKCB
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human PKCbII that contains threonine 641. The sequence is conserved in mouse and rat.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human