Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PKC delta (Ser664), Polyclonal, Invitrogen™

Catalog No. PIPA126722
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA126722 50 μL
1 options

Catalog No. PIPA126722

Supplier: Thermo Scientific PA126722

Rabbit Polyclonal Antibody

PA1-26722 detects PKC delta (phospho S664) from human samples.PA1-26722 has been successfully used in Western blot procedures.The PA1-26722 immunogen is a synthetic phosphopeptide corresponding to a portion of human PKC delta containing serine 664.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKC delta pSer664
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene PRKCD
Gene Accession No. Q05655
Gene Symbols PRKCD
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to a portion of human PKC delta containing serine 664.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5580
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.