Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PKC gamma (Thr514) Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen 44956

Catalog No. 44-956


Add to cart

Description

Description

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
Specifications

Specifications

Phospho-PKC gamma (Thr514)
Polyclonal
Dulbecco′s PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
P05129, P63318
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Rabbit
IgG
100 μL
-20°C
Primary
18752, 5582
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PRKCG
Liquid
PRKCG
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human PKC-gamma that contains threonine 514
Purified
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit