Learn More
Phospho-PP2A Mouse anti-Vertebrates, Clone: 4B10, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05547
Catalog No. 05-547-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-phospho-PP2A Antibody, C subunit (Tyr307), clone 4B10 detects level of phospho-PP2A and has been published and validated for use in ELISA.
Specifications
Specifications
phospho-PP2A
4B10
mouse ascites containing 30% glycerol and 0.05% azide
Ascites
Murine
IgG1
100μL
Primary
Vertebrate
ELISA
Unconjugated
NM_004156
PPP2CA, PP2CA, PP2A-alpha, PP2Ac, RP-C
Peptide corresponding to residues 302-309 of human PP2A; recognizes phosphorylated PP2A on tyrosine 307.
Ascites
−20°C for 24 months.
Monoclonal