Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-PPARγ (Ser82) Rabbit anti-Human, Mouse, Rat, Clone: AW504, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04816
Description
Use Anti-phospho-PPAR gamma antibody, clone AW504 (rabbit monoclonal antibody) validated in western blotting to detect phospho-PPARγ (Ser82) also known as Nuclear receptor subfamily 1 group C member 3 and PPAR gamma.Specifications
phospho-PPARγ (Ser82) | |
Monoclonal | |
Unconjugated | |
NP_005028.4 | |
KLH-conjugated, synthetic peptide containing the sequence PApSPP in which pS corresponds to phosphoserine at residue 82 of human PPARγ. | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
AW504 | |
Cultured supernantant containing 0.05% sodium azide. | |
Rabbit | |
100 μL | |
CIMT1, NR1C3, OTTHUMP00000185030, OTTHUMP00000185033, OTTHUMP00000185037, PPAR-gamma, PPARG1, PPARG2 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only