Learn More
Phospho-PYK2 (Tyr579), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA126673
Description
This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. At least four transcript variants encoding four different isoforms have been found for this gene, but the full-length natures of only two of them have been determined.
PA1-26673 detects PYK2 (phospho Y579) from human samples. PA1-26673 is expected to cross react with mouse (100% conserved) and rat (100% conserved) due to sequence homology.PA1-26673 has been successfully used in Western blot procedures.The PA1-26673 immunogen is a synthetic phosphopeptide corresponding to a portion of human Pyk 2 containing tyrosine 579. The sequence is conserved in mouse and rat.
Specifications
Phospho-PYK2 pTyr579 | |
Polyclonal | |
PTK2B | |
FADK, FAK, FAK1, FRNK, pp125FAK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2185 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Purified |
Western Blot | |
Unconjugated | |
Q14289, Q9QVP9, P70600 | |
PTK2B | |
Synthetic phosphopeptide corresponding to a portion of human Pyk 2 containing tyrosine 579. The sequence is conserved in mouse and rat. | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.