Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Rb (Ser612) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512680

Catalog No. PIPA512680


Add to cart

Description

Description

Retinoblastoma (RB) is an embryonic maligt neoplasm of retinal origin. It almost always presents in early childhood and is often bilateral. Spontaneous regression ( ft.cure ft.) occurs in some cases.[supplied by OMIM].
Specifications

Specifications

Phospho-Rb (Ser612)
Polyclonal
Unconjugated
RB1
Liquid
RB1
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S612 of human RB
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 0.09% sodium azide
P06400
Retinoblastoma-associated protein; PP110; P105-RB; RB; RB1;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5925
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit