Learn More
Phospho-Rb (Ser788) Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA512682
Catalog No. PIPA512682
Add to cart
Description
Description
RB1 likely acts as a regulator of other genes. It forms a complex with adenovirus E1A and with SV40 large T antigen, acts as a tumor suppressor, and may bind and modulate functionally certain cellular proteins with which T and E1A compete for pocket binding. RB1 is a potent inhibitor of E2F-mediated trans-activation, and also recruits and targets histone methyltransferase SUV39H1 leading to epigenetic transcriptional repression. This protein inhibits the intrinsic kinase activity of TAF1. Defects in RB1 are the cause of childhood cancer retinoblastoma (RB), a congenital maligt tumor that arises from the nuclear layers of the retina. Defects in RB1 are also a cause of bladder cancer and osteogenic sarcoma.
Specifications
Specifications
Phospho-Rb (Ser788)
Polyclonal
Unconjugated
RB1
Liquid
RB1
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S788 of human Rb
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Dot blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
P06400
Retinoblastoma-associated protein; PP110; P105-RB; RB; RB1;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5925