Learn More
Phospho-RSK1 (Ser380), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124771
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
PA1-24771 detects RSK1 p90 (phospho S380) from human and mouse samples. PA1-24771 is expected to cross react with rat (100% conserved) due to sequence homology. Cross reactivity with RSK2 is expected (85% conserved) based on peptide competition.PA1-24771 has been successfully used in Western blot procedures.The PA1-24771 immunogen is a synthetic phosphopeptide corresponding to a portion of human RSK1 containing serine 380.
Specifications
Phospho-RSK1 p90 pSer380 | |
Polyclonal | |
RPS6KA1 | |
RP11-492M19.2, HU-1, MAPKAPK1A, RSK, RSK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6195 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
Q15418, P18653, Q63531 | |
RPS6KA1 | |
Synthetic phosphopeptide corresponding to a portion of human RSK1 containing serine 380. | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.