Learn More
Phospho-RSK1 (Ser380) Rabbit anti-Chicken, Human, Mouse, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen 44928G
Catalog No. 44-928-G
Add to cart
Description
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
Phospho-RSK1 (Ser380)
Polyclonal
Dulbecco′s PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3
Q15418, P18653
HU-1, MAPKAPK1A, RSK, RSK1, 90 kDa ribosomal protein S6 kinase 1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, RSK-1, S6K-alpha 1, S6K-alpha-1, dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1), p90-RSK 1, p90RSK1, p90S6K, r
Rabbit
IgG
100 μL
-20°C
Primary
20111, 6195
Western Blot
Unconjugated
RPS6KA1
Liquid
RPS6KA1
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human RSK1 that contains serine 380. The sequence is conserved in mouse and rat.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Chicken, Human, Mouse