Learn More
Phospho-Ser/Thr-Pro MPM-3 Mouse anti-Chicken, Human, Mouse, Rat, Clone: CC-3, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05918
Catalog No. 05-918-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-phospho-Ser/Thr-Pro MPM-3 Antibody, clone CC-3 is a Mouse Monoclonal Antibody for detection of phospho-Ser/Thr-Pro MPM-3 also known as Mitotic protein #3 and has been tested in immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
phospho-Ser/Thr-Pro MPM-3
CC-3
Ascites
Cell homogenates from pharyngeal chick embryos
100μL
Primary
Chicken, Human, Murine, Rat
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
Murine
Ascites
−20°C for 24 months.
Monoclonal