Learn More
Phospho-(Ser10) acetyl (Lys14) Histone H3 Mouse anti-Human, Clone: 10H10.2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051315
Catalog No. 05-131-5MI
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-phospho (Ser10) acetyl (Lys14) Histone H3 Antibody, clone 10H10.2 (Mouse Monoclonal Antibody) validated in western blotting to detect phospho (Ser10) acetyl (Lys14) Histone H3 also known as H3S10PK14Ac.
Specifications
Specifications
phospho (Ser10) acetyl (Lys14) Histone H3
Monoclonal
Unconjugated
NP_002098.1
Mouse
Protein G purfied
Primary
2-8°C for one year
IgG1 κ
Western Blot
10H10.2
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
HIST3H3, H3FT, H3t, H3T, H3/g, H3.4, H3/t
KLH-conjugated linear peptide corresponding to Histone H3 phosphorylated at Ser10 and acetylated at Lys14.
100 μg
Human
Purified