Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-AANAT (Ser206) Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1763

Catalog No. PA1763

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Pineal gland serotonin N-acetyltransferase (arylalkylamine N-acetyltransferase, AA-NAT) is an ~23kDa protein which is the rate-limiting enzyme in melatonin synthesis. Melatonin serves as the hormonal signal of the daily light/dark cycle, or body clock. The nocturnal oscillation in enzyme activity ranges by 10- to 100-fold depending on species among vertebrates. Studies have shown that AA-NAT is regulated by cAMP at both the mRNA and protein levels. AA-NAT has putative protein kinase A (PKA) sites at Thr29 and Ser206. Phosphorylation of AA-NAT at these residues might play a role in protein inactivation via proteasome-dependent degradation.

PA1-763 detects phospho serotonin N-acetyltransferase (AA-NAT) from rat tissues.PA1-763 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~23kDa protein representing phosphorylated AA-NAT from rat pineal gland whole protein extract. PA1-763 immunizing phosphopeptide (Cat. # PEP-066) corresponds to carboxy-terminal amino acid residues 198-208 of rat AA-NAT. Total anti-carboxy-terminal AA-NAT-specific IgG (Cat. # PA1-762) was affinity purified on resin-coupled non-phosphopeptide (Cat. # PEP-065). Phospho-specific carboxy-terminal IgG (Cat. # PA1-763) was subsequently isolated on resin-coupled phosphopeptide (Cat. # PEP-066). This sequence is completely conserved between rat and mouse AA-NAT protein.
Specifications

Specifications

Phospho-AANAT (Ser206)
Polyclonal
Unconjugated
Aanat
Liquid
AANAT
Synthetic peptide: C(195) H T F L R R N S(p) G C(205)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q64666
Phospho-Serotonin N-Acetyltransferase
Rabbit
IgG
50 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
25120
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit