Learn More
Phospho-SMC1 (Ser957) Mouse anti-Human, Mouse, Clone: 5D11G5, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05786
Catalog No. 05-786-MI
Add to cart
Description
Description
Detect phospho-SMC1 (Ser957) with Anti-phospho-SMC1 (Ser957) Antibody, clone 5D11G5 (Mouse Monoclonal Antibody), that has been shown to work in western blotting.
Specifications
Specifications
phospho-SMC1 (Ser957)
5D11G5
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG1
200μg
Primary
Human, Murine
Western Blot
Unconjugated
NM_006306
SMC1A, SMCB, SMC1alpha, DXS423E, DKFZp686L19178, SMC1, SMC1L1, Smcb, OTTHUMP00000061876, KIAA0178, SB1.8, MGC138332, CDLS2, Sb1.8
KLH-conjugated, synthetic peptide corresponding to amino acids 951-962 (SQEEGS[pS]QGEDS) of human SMC1
Protein G purfied
−20°C for 24 months.
Monoclonal