Learn More
Phospho-Src family (Tyr416) Rabbit anti-Mouse, Vertebrates, Clone: 2N8, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05857
Catalog No. 05-857-MI
Add to cart
Description
Description
This Anti-phospho-Src family (Tyr416) Antibody, clone 2N8 is validated for use in western blotting for the detection of phospho-Src family (Tyr416).
Specifications
Specifications
phospho-Src family (Tyr416)
2N8
NM_005417
Rabbit
IgG
100μL
Primary
SRC, c-src, p60-Src, SRC1, c-SRC, ASV, pp60c-src
Western Blot
Unconjugated
Purified
synthetic peptide containing the sequence ...NE[pY]TA
Protein A purfied
−20°C for 24 months
Monoclonal
Murine, Vertebrate
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    01-31-2021