Learn More
Phospho-Src (Tyr416) Mouse anti-Human, Mouse, Clone: 9A6, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05677
Catalog No. 05-677-MI
Add to cart
Description
Description
Detect phospho-Src (Tyr416) using this Anti-phospho-Src (Tyr416) Antibody, clone 9A6 validated for use in western blotting and immunoprecipitation.
Specifications
Specifications
phospho-Src (Tyr416)
9A6
Purified mouse monoclonal IgG3 lyophilized from 1 mL 2x PBS / 0.09 % Na-azide / PEG and Sucrose. Reconstitute with 1 mL H2O (15 min, RT). Lyophilized. Store at −20deg;C.
Purified
Murine
IgG3
100μg
Primary
Human, Murine
Immunoprecipitation, Western Blot
Unconjugated
NM_005417
SRC, c-src, p60-Src, SRC1, c-SRC, ASV, pp60c-src
KLH-conjugated, synthetic peptide containing phosphotyrosine in the sequence context corresponding to tyrosine 416 of avian Src. Clone 9A6.
Thiophilic adsoption and size exclusion chromatography
Stable for 1 year at from date of receipt.
Monoclonal