Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-STAT1 (Ser 727) Mouse anti-Human, Clone: 12C5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04478
Description
Anti-phospho-STAT1 (Ser 727) Antibody, clone 12C5 is a Mouse Monoclonal Antibody for detection of phospho-STAT1 (Ser 727) also known as signal transducer and activator of transcription 1 and has been tested in ELISA and western blotting.Specifications
phospho-STAT1 (Ser 727) | |
Monoclonal | |
Unconjugated | |
NM_007315.2; NM_139266.1 | |
Mouse | |
Subsequent thiophilic adsorption and size exclusion chromatography | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
ELISA, Western Blot | |
12C5 | |
In 1 mL of PBS containing 0.09% sodium azide, PEG, sucrose, and 50% glycerol. | |
STAT1, ISGF-3, STAT91, DKFZp686B04100 | |
KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Serine727 on phosphorylated STAT1 | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only