Learn More
Phospho-STAT6 (Tyr641) Mouse anti-Human, Mouse, Rat, Clone: 16E12, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05590
Catalog No. 05-590-MI
Add to cart
Description
Description
Detect phospho-STAT6 (Tyr641) also known as signal transducer and activator of transcription 6 with Anti-phospho-STAT6 (Tyr641) Antibody, clone 16E12 (mouse monoclonal antibody), that has been demonstrated to work in western blotting.
Specifications
Specifications
phospho-STAT6 (Tyr641)
16E12.
100μg of lyophilized thiophilic and size exclusion chromatography purified mouse IgG1 from 1mL 2x PBS / 0.09 % Na-azide / PEG and Sucrose. Reconstitute with 100μL H2O (15 min, RT) for a final concentration of 1mg/mL. Store at −20°C.
Purified
Murine
IgG1
100μg
Primary
Human, Murine, Rat
Western Blot
Unconjugated
NM_003153
STAT6, STAT6C, D12S1644, STAT6B, IL-4-STAT
Peptide corresponding to residues of human STAT6 (Tyr641); recognizes STAT6 phosphorylated on Tyrosine 641.
Thiophilic adsoption and size exclusion chromatography
−20°C for 24 months.
Monoclonal