Learn More
Phospho-Synaptotagmin 1 (Ser309) Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14641
Catalog No. PA14641
Add to cart
Description
Description
This antibody is predicted to react with bovine, canine, chicken, human, mouse, non-human primate and zebrafish based on 100% sequence homology. This antibody is specific for the ~60 kDa to 62 kDa Synaptotagmin protein phosphorylated at Ser309 in Western blots of rat brain lysates.
Synaptotagmin 1 is a synaptic vesicle membrane glycoprotein that is widely expressed throughout the CNS and is generally thought to act as the Ca2+ sensor in the regulation of exocytosis and neurotransmitter release. Studies indicate that synaptotagmin’s Ca2+ mediated binding of SNAP25 is essential for the Ca2+ dependent triggering of membrane. It has been demonstrated that discrete residues within the c(2)b binding domain of synaptotagmin 1 independently specify endocytic rate and synaptic vesicle size. Synaptotagmin 1 may have a regulatory role in the membrane interactions during trafficking of synaptic vesicles at the active zone of the synapse. Further, Synaptotagmin 1 binds acidic phospholipids with a specificity that requires the presence of both an acidic head group and a diacyl backbone. A Ca(2+)-dependent interaction between synaptotagmin and putative receptors for activated protein kinase C has been reported. Synaptotagmin 1 can also bind to at least three additional proteins in a Ca(2+)-independent manner; these are neurexins, syntaxin and AP2. Diseases associated with the dysfunction of Synaptotagmin 1 include food-borne botulism and infant botulism.
Specifications
Specifications
Phospho-Synaptotagmin 1 (Ser309)
Polyclonal
Unconjugated
Syt1
Liquid
SYT1
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser309 of rat SYT1 conjugated to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Rat
Dot Blot, Immunohistochemistry, Western Blot
0.1 mg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P21707
SVP65, SYT
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25716