Learn More
Phospho-alpha Synuclein (Ser129) Rabbit anti-Rat, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen 482700
Description
48-2700 is predicted to react with bovine, canine, human, mouse and non-human primates based on 100% sequence homology. This product contains enough material to conduct 10 mini-Western blots.
Alpha-Synuclein belongs to the Synuclein family, which includes beta and gamma Synuclein, and is predomitly expressed in neurons, and concentrated at synaptic terminals. Synucleins may be involved in the regulation of dopamine release and transport and the modulation of synaptic vesicle function. Alpha-Synuclein is a small, presynaptic protein that is the major nonamyloid component of the pathological inclusions characteristic of a wide range of neurodegenerative disorders, collectively known as synucleinopathies. Alpha-Synuclein is involved in the formation of SNARE complexes, and most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson ft.s disease (PD) and other neurodegenerative disorders. Mutation of the alpha-synuclein gene is associated with familial forms of PD. Alpha-Synuclein is known to reduce the fibrillization of the microtubule-associated protein, tau, to bind various targets such as 14-3-3, protein kinase C, synphilin-1, Elk and Tat-binding protein 1. The alpha and beta synucleins are found primarily in brain tissue where they are seen mainly in neuronal cytosol and presynaptic terminals. The antibody specific for alpha-synuclein does not cross react with beta-synuclein and will be useful for the confirmation of Parkinson ft.s disease and Lewy body dementia and other studies for the localization and detection of alpha-synuclein. Four alternatively spliced transcripts that encode two different isoforms of the alpha-synuclein protein have been identified.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Phospho-alpha Synuclein (Ser129) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P37377 | |
NACP, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, SYN, SYUA, alpha-synuclein, synuclein, alpha | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
29219 |
Western Blot | |
Unconjugated | |
Snca | |
Liquid | |
Snca | |
Phosphopeptide corresponding to amino acid residues surrounding the phosphoserine 129 of -alpha-synuclein. | |
Affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Rat |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.