Learn More
Phospho-TAK1 (Ser412) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061425
Catalog No. 06-142-5MI
Add to cart
Description
Description
Detect phospho-TAK1 (Ser412) using this Anti-phospho-TAK1 (Ser412) Antibody validated for use in western blotting.
Specifications
Specifications
phospho-TAK1 (Ser412)
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human, Murine
Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
MAP3K7; TAK1
KLH-conjugated linear peptide corresponding to human TAK1 phosphorylated at Ser412.
200μL
Primary
NP_663306
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    01-31-2021