Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-TAOK2 (Ser181) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA14643
Description
This antibody is predicted to react with zebrafish, Xenopus, finch and canine based on 100% sequence homology. This antibody is specific for the ∽120 kDa TAO2 phosphorylated at Ser181 in Western blots of rat hippocampal lysate.
Phospho-TAOK2 (Ser181) Polyclonal antibody specifically detects Phospho-TAOK2 (Ser181) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-TAOK2 (Ser181) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q6ZQ29, Q9JLS3, Q9UL54 | |
TAOK2 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser181 of human TAOK2 conjugated to KLH | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TAOK2 | |
1110033K02Rik; B230344N16; hKFC-C; Kiaa0881; Kinase from chicken homolog C; MAP3K17; mKIAA0881; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; prostate-derived sterile 20-like kinase 1; PSK; PSK1; PSK-1; PSK1-BETA; serine/threonine protein kinase TAO2; serine/threonine-protein kinase TAO2; serine-threonine kinase; tao; TAO kinase 2; TAO kinase 2TAO kinase 2; TAO1; TAO2; TAOK2; Thousand and one amino acid protein 2; thousand and one amino acid protein kinase 2; UNQ2971/PRO7431 | |
Rabbit | |
Affinity chromatography | |
RUO | |
381921, 64666, 9344 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction