Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-TSC2 S664, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of TSC2.

Manufacturer:  Abnova Corporation PAB8108

Catalog No. 89-024-847


Add to cart

Description

Description

Mutations in this gene lead to tuberous sclerosis complex. Its gene product is believed to be a tumor suppressor and is able to stimulate specific GTPases. The protein associates with hamartin in a cytosolic complex, possibly acting as a chaperone for hamartin. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Specifications

phospho-TSC2 S664
Unconjugated
ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user.
TSC2
FLJ43106|LAM|TSC4
Rabbit
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Dot Blot, ELISA
Rabbit polyclonal antibody raised against synthetic phosphopeptide of TSC2.
In PBS (0.09% sodium azide)
Liquid
TSC2
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S664 of human TSC2.
RUO
Primary
7249
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit