Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-YES1/SRC (Tyr537, Tyr530) Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512698
Description
This antibody is predicted to react with chicken, mouse, rat and zebrafish based on sequence homology.
This protein has tyrosine kinase activity and belongs to the src family of proteins. The YES1 gene is the cellular homolog of the Yamaguchi sarcoma virus oncogene.Specifications
Phospho-YES1/SRC (Tyr537, Tyr530) | |
Polyclonal | |
SRC | |
Proto-oncogene tyrosine-protein kinase Yes; p61-Yes; c-Yes; YES1; YES; Proto-oncogene tyrosine-protein kinase Src; EC 2.7.10.2; pp60c-src; p60-Src; c-Src | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6714, 7525 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
P07947, P12931 | |
SRC, YES1 | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y530 of human SRC | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction