Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
MilliporeSigma™ anti-PhosphoDetect Paxillin (pSer178), Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST106950UG
Catalog No. ST106950UG
Description
Description
Specifically detects PhosphoDetect™ Paxillin (pSer178) Clone: in Human, Mouse samples, and it is validated for Immuno Blotting
Paxillin is a focal adhesion adaptor protein associated with th establishment of focal adhesions, cell signaling via integrins, and cell motility. The N-terminus of paxillin contains a proline-rich region for SH3 and SH2 domain binding. The N-terminus also contains five copies of the LD motif for association of actin binding proteins. The C-terminus contains LIM domains and the focal adhesion targeting sequence. Paxillin is believed to function as an adaptor protein that facilitates assembly of multi-protein complexes and coordinates signaling via integrins. Tyrosine and serine phosphorylation of paxillin is associated with the establishment of cytoskeletal membrane attachments. Phosphorylation by JNK at Ser178 is necessary for normal cell motility.
Specifications
Specifications
PhosphoDetect™ Paxillin (pSer178)
Unconjugated
purified
a synthetic phosphopeptide corresponding to amino acids surrounding the Ser178 phosphorylation site of human paxillin
50μg
2°C to 8°C
Polyclonal
Human, Murine
Immuno Blotting, Immunoblot
In Tris-citrate/phosphate, pH 7.0-8.0.
Rabbit
IgG
RUO
Primary
Recognizes the ∽68kDa paxillin protein phosphorylated at Ser178.