Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phosphodiesterase 4D, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131131

Catalog No. NBP131131


Add to cart

Description

Description

Phosphodiesterase 4D Polyclonal antibody specifically detects Phosphodiesterase 4D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Phosphodiesterase 4D
0.3mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
PDE4D
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PDE4D. The exact sequence is proprietary.
91 kDa
0.1 ml
Lipid and Metabolism, Signal Transduction
Primary
5144
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E3), cAMP-specific 3'-5'-cyclic phosphodiesterase 4D, cAMP-specific phosphodiesterase PDE4D6, DKFZp686M11213, Drosophila), dunce-like phosphodiesterase E3, EC 3.1.4, FLJ97311, HSPDE4D, phosphodiesterase 4D, cAMP-specific, phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog
Rabbit
IgG
Antigen Affinity-purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only