Learn More
PGK1 Mouse anti-Yeast, Clone: 22C5D8, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen 459250
Catalog No. 45-925-0
Add to cart
Description
Description
The protein encoded by this gene is a glycolytic enzyme that catalyzes the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate. The encoded protein may also act as a cofactor for polymerase alpha. This gene lies on the X-chromosome, while a related pseudogene also has been found on the X-chromosome and another on chromosome 19.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
PGK1
Monoclonal
1 mg/mL
HEPES buffered saline with 0.02% sodium azide
Liquid
PGK1
Yeast pGK (3-Phosphoglyceric Phosphokinase), EC 2.7.2.3
Ammonium Sulfate Precipitation
RUO
Antibody
Monoclonal
Yeast
Immunocytochemistry, Immunofluorescence, Western Blot
22C5D8
Unconjugated
PGK1
MIG10, PGKA, PRP 2, cell migration-inducing gene 10 protein, primer recognition protein 2
Mouse
IgG1, kappa
250 μg
4° C
Primary
850370