Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G12A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522204

Catalog No. PIPA522204


Add to cart

Description

Description

PA5-22204 targets Phospholipase A2 XIIA in WB applications and shows reactivity with Human samples. The PA5-22204 immunogen is a synthetic peptide corresponding to a region within amino acids 22 and 115 of PLA2G12A (Uniprot IDNo.Q9BZM1). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

PLA2G12A
Polyclonal
Unconjugated
PLA2G12A
Liquid
PLA2G12A
Synthetic peptide corresponding to a region within amino acids 22 and 115 of PLA2G12A (Uniprot IDNo.Q9BZM1)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.3 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9BZM1
FKSG38, GXII, PLA2G12, ROSSY, GXII sPLA2, group XII secreted phospholipase A2, group XIIA secreted phospholipase A2, group XIIA secretory phospholipase A2, phosphatidylcholine 2-acylhydrolase 12A, sPLA2-XII
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
81579
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit