Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCB1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138470

Catalog No. 01-673-863

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is brain tissue.

Phosphoinositide-specific phospholipase C (PLC) plays a critical role in the initiation of receptor mediated signal transduction through the generation of the two second messengers, inositol 1,4,5-triphosphate and diacylglycerol from phosphatidylinositol 4,5 bisphosphate. There are many mammalian PLC isozymes, including PLC β1, PLC β2, PLC β3, PLC β4, PLCγ1, PLCγ2, PLC δ1, PLC δ2 and PLC ε with molecular weights ranging from 85 to 150kDa.
Specifications

Specifications

PLCB1
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.4
Q9NQ66
EIEE12, PI-PLC, PLC-154, PLC-I, PLC154, PLCB1A, PLCB1B, 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1, 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta 1, 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1, PLC-beta-1, in
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23236
Immunohistochemistry (Paraffin)
Unconjugated
PLCB1
Liquid
PLCB1
Synthetic peptide corresponding to C-terminus of human PLC beta 1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit