Learn More
PLSCR1 Mouse anti-Human, Clone: 3B4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519636
Catalog No. PIMA519636
Add to cart
Description
Description
Peptide Sequence: MDKQNSQMNA SHPETNLPVG YPPQYPPTAF QGPPGYSGYP GPQVSYPPPP AGHSGPGPAG FPVPNQPVYN QPVYNQPVGA AGVPWMPAPQ PPLNCPPGLE YLSQIDQILI HQQIKLLEVL TGFETNNKYE IKNSFGQRVY FAAEDTDCCT RNCCGPSRPF TLRIIDNMGQ EVITLERPLR CSSCCCPCCL QEIEIQAPPG VPIGYVIQTW HPCLPKFTIQ NEKREDVLKI SGPCVVCSCC GDVDFEIKSL DEQCVVGKIS KHWTGILREA FTDADNFGIQ FPLDLDVKMK AVMIGACFLI DFMFFESTGS QEQKSGVW
Scramblase-1 is a calcium-binding protein that can be induced by interferon and growth factors (e.g., epidermal growth factor, EGF). Depending on its palmitoylation state, scramblase-1 either inserts into the plasma membrane or binds DNA in the nucleus. Scramblase-1 is a substrate for Src kinases.
Specifications
Specifications
PLSCR1
Monoclonal
0.2 mg/mL
PBS with no preservative; pH 7.4
O15162
MMTRA1B, PL scramblase 1, ca(2+)-dependent phospholipid scramblase 1, erythrocyte phospholipid scramblase
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5359
ELISA, Immunoprecipitation
3B4
Unconjugated
PLSCR1
Liquid
PLSCR1
PLSCR1 (AAH32718, 1 a.a. ∽ 318 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human