Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phosphorylase B, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128838

Catalog No. NBP128838

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Phosphorylase B
Unconjugated
PBS with 0.09% Sodium Azide
Purified
PHKB
Native, from muscle
Protein G purified
RUO
Primary
5257
Rabbit
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P12798
DKFZp781E15103, FLJ41698, phosphorylase b kinase regulatory subunit beta, phosphorylase kinase beta-subunit, Phosphorylase kinase subunit beta, phosphorylase kinase, beta
Sheep
IgG
0.5 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
PHOSPHORYLASE B
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only