Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-serine Mouse anti-Vertebrates, Clone: 4A4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051000MG
Description
This Anti-Phosphoserine Antibody, clone 4A4 is validated for use in western blot, immunofluoresence, flow cytometry, immunohistochemistry (paraffin), ELISA for the detection of Phosphoserine.Specifications
Phosphoserine | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein G-Sepharose™ chromatography | |
Primary | |
2 years at -20°C from date of shipment. For maximum recovery of the product, centrifuge the original vial prior to removing the cap. If the product has accidentally been frozen and thawed, spin it at 13,000 x g for 10 minutes at 2-8°C. | |
IgG1 |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
4A4 | |
1 mL of PBS | |
Phosphoserine coupled to KLH | |
1 mg | |
Vertebrates | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only