Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phosphoserine phosphatase, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128916

Catalog No. NBP128916


Add to cart

Description

Description

Phosphoserine phosphatase Polyclonal antibody specifically detects Phosphoserine phosphatase in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

Phosphoserine phosphatase
0.5 mg/mL
Western Blot 0.05 - 1 ug/ml, Peptide ELISA Detection limit 1:32000
NP_004568.2
L-3-phosphoserine phosphatase, O-phosphoserine phosphohydrolase, phosphoserine phosphatase, PSPase, PSPEC 3.1.3.3
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human and Mouse. Expected from sequence similarity: Rat.
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
PSPH
Peptide with sequence C-RQQVKDNAKWYITD corresponding to C-Terminus according to NP_004568.2.
Immunogen affinity purified
RUO
Primary
5723
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only