Learn More
Phosphoserine/threonine/tyrosine Mouse anti-Chemical, Clone: SPM101, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA138450
Catalog No. 01-672-764
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-38450 detects phosphotyrosine, phosphoserine, and phosphothreonine in WB and ELISA applications.
There are three amino acids that are typically phosphorylated in eukaryotes: serine, threonine, and tyrosine. These phosphoric acid esters can be used as an identifier in the analysis of peptides, proteins, and enzymes. Antibodies that specifically recognize phosphorylated residues have proved to be invaluable to the study of biochemical pathways in which they function.
Specifications
Specifications
Phosphoserine/threonine/tyrosine
Monoclonal
Unconjugated
Liquid
Mouse
IgG1
1 mL
-20°C
Primary
Chemical
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
SPM101
ascites diluted in PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Phospho-serine; Phospho-threonine; Phospho-tyrosine; pSer; pTyr; pThr; serine; threonine; tyrosine
Phosphoserine, phosphothreonine, and phosphotyrosine conjugated to protein carriers.
Purified
RUO
Antibody
Monoclonal