Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phosphotyrosine Ab-1, Clone: PY20, Thermo Scientific Lab Vision

Mouse Monoclonal Antibody

Specifications

Antigen Phosphotyrosine Ab-1
Format Purified
Host Species Murine
Immunogen Phosphotyrosine conjugated to a carrier protein.
Isotype IgG2b
View More Specs

Products 7
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
MS445P
View Documents
Epredia
MS445P
200μg/mL Unconjugated 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS445P0
View Documents
Epredia
MS445P0
200μg/mL Unconjugated 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS445P1
View Documents
Epredia
MS445P1
200μg/mL Unconjugated 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS445P1ABX
View Documents
Epredia
MS445P1ABX
1mg/mL Unconjugated 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS445PCS
View Documents
Epredia
MS445PCS
n/a Unconjugated 5 Slides This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS445PABX
View Documents
Epredia
MS445PABX
1mg/mL Unconjugated 200μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS445R7
View Documents
Epredia
MS445R7
n/a Unconjugated 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Protein phosphorylation is a fundamental event in the regulation of a large number of intracellular processes. Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be autophosphorylated kinases or certain cellular protein substrates. Tyrosine-phosphorylated proteins are involved in signal transduction and in the regulation of cell proliferation. Antibody to phosphotyrosine provides an excellent tool for the detection, characterization, and purification of phosphotyrosine containing proteins.

Host Species: Mouse
Clone: PY20
Isotype: IgG2b
Cross Reactivity: All species.
Immunogen: Phosphotyrosine conjugated to a carrier protein.
Positive Control: MCF-7, MDA-231, T47-D cells or breast CA.
Cellular Localization: Cell surface and cytoplasmic

Ab-1 shows no cross-reaction with other phosphoamino acids and is excellent for multiple applications including staining of formalin/paraffin tissues.

Immunofluorescence, Immunohistochemistry (Formalin/Paraffin), Immunoprecipitation, Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

Phosphotyrosine Ab-1
Murine
IgG2b
Primary
Purified
Phosphotyrosine conjugated to a carrier protein.
Monoclonal
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit