Learn More
Phospho-tyrosine Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06427
Catalog No. 06-427-MI
Add to cart
Description
Description
Anti-Phosphotyrosine Antibody detects level of Phosphotyrosine and has been published and validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
Phosphotyrosine
Unconjugated
Affinity Purified
In order to produce broad spectrum polyclonal phosphotyrosine antibodies, rabbits were immunized with three phosphorylated immunogens: (1) phosphotyrosine covalently linked to KLH; (2) the c-Src carboxyl terminal regulatory phosphopeptide (TS-T-E-P-Q-pY-Q
100μL
Primary
Human
Immunoprecipitation, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal