Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine Mouse, Agarose, Clone: PY69, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610015

Catalog No. BDB610015


Add to cart

Description

Description

Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be autophosphorylated kinases or certain cellular protein substrates that are regulated

Host Species: Mouse
Clone: PY69
Isotype: IgG2b
Species Reactivity: Human

Immunofluorescence, Immunohistochemistry, Immunoprecipitation

Specifications

Specifications

Phosphotyrosine
PY69
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
1mL
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
Immunoprecipitation
Agarose
Affinity Purified
IgG2a
RUO
Store undiluted at 4°C
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit