Learn More
Phosphotyrosine Mouse anti-Chemical, Rhodamine, Clone: pY20, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112448
Catalog No. 01-674-315
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
Specifications
Specifications
Phosphotyrosine
Monoclonal
1 mg/mL
PBS with 50% glycerol and 3mM sodium azide; pH 7.5
pTyr; pY; Phosphotyrosine
Phosphotyrosine coupled to a carrier protein
Purified
RUO
Antibody
Monoclonal
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
PY20
Rhodamine
Liquid
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical