Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHPT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512294

Catalog No. PIPA512294


Add to cart

Description

Description

PHPT1 is an EDTA-insensitive phosphohistidine phosphatase that catalyzes the dephosphorylation of phosphopeptide I.
Specifications

Specifications

PHPT1
Polyclonal
PBS with 0.09% sodium azide
Q9NRX4
14 kDa phosphohistidine phosphatase;Phosphohistidine phosphatase 1;PHPT1;PHP14
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29085
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PHPT1
Liquid
PHPT1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PHPT1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit