Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHPT1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512295
Description
PHPT1 is an EDTA-insensitive phosphohistidine phosphatase that catalyzes the dephosphorylation of phosphopeptide I.Specifications
PHPT1 | |
Polyclonal | |
PHPT1 | |
14 kDa phosphohistidine phosphatase; Phosphohistidine phosphatase 1; PHPT1; PHP14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29085 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9NRX4 | |
PHPT1 | |
KLH conjugated synthetic peptide between 88-117 amino acids from the C-terminal region of human PHPT1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction