Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHTF2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523940

Catalog No. PIPA523940

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken based on sequence homology.

Specifications

Specifications

PHTF2
Polyclonal
PBS with 0.09% sodium azide
Q8N3S3, Q8CB19
PHTF2
KLH conjugated synthetic peptide between 420-448 amino acids from the central region of human PHTF2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
Unconjugated
PHTF2
Liquid
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57157, 68770
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit