Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phycoerythrin B Rabbit anti-Tag, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128741

Catalog No. PIPA128741


Add to cart

Description

Description

PA1-28741 detects Phycoerythrin B from tagged samples. PA1-28741 has been successfully used in ELISA, Western blot applications. The PA1-28741 immunogen is: B-Phycoerythrin [Porphyridium cruentum] Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Phycoerythrin B
Polyclonal
0.02M potassium phosphate with 0.15M NaCl and no preservative; pH 7.2
B-Phycoerythrin
B-Phycoerythrin [Porphyridium cruentum]
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Tag
ELISA, Western Blot
Unconjugated
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit